Desiccators with PROTEFAN safety coating
Lieferant: Witeg
WITG5.065.300EA 626.66 EUR
WITG5.065.300 WITG5.065.200
Desiccators with PROTEFAN safety coating
Exsikkatoren Exsikkatoren, Glas
Desiccators are made of borosilicate glass, with safety PROTEFAN coating as implosion and splinter protection.