Centrifuge tubes, conical bottom
Lieferant: Witeg
WITG4.945.005EA 17.2 EUR
WITG4.945.005
Centrifuge tubes, conical bottom
Röhrchen Zentrifugenröhrchen
DURAN®, borosilicate glass.
  • Heavy wall
  • Ungraduated
  • Very good chemical resistance
  • Very good temperature resistance
Conical bottom, with out rim