Condensers, Graham
Lieferant: Witeg
WITG1.733.400EA 95.39 EUR
WITG1.733.400 WITG1.734.400 WITG1.738.250 WITG1.732.160 WITG1.737.250 WITG1.733.250 WITG1.734.250
Condensers, Graham
Kühler
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Coil type, Graham pattern, suitable for either reflux or distillation work. Water connection with glass olives.