Distilling bridges
Lieferant: Witeg
WITG1.205.300EA 119.27 EUR
WITG1.205.300 WITG1.205.301 WITG1.205.260 WITG1.205.150
Distilling bridges
Destillationsbauteile Destillierbrücken
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.