Stopcock, three-way, acc. to Czako
Lieferant: Witeg
WITG0.268.001EA 60.85 EUR
WITG0.268.001
Stopcock, three-way, acc. to Czako
Hähne
Stopcocks made of borosilicate glass.
  • With capillary side arms
  • Solid glass key
  • Good chemical and temperature resistance
Standards: DIN 12563