Hose connections
Lieferant: Witeg
WITG0.186.004EA 5.94 EUR
WITG0.186.004 WITG0.185.007 WITG0.185.008 WITG0.186.014 WITG0.185.014 WITG0.185.004 WITG0.185.005 WITG0.185.006
Hose connections
Fittings und Verbinder
Hose connections, bent or straight, made of polypropylene (PP) or PFA, with silicone rubber liners.