Sieve standards, polydisperse particle standard
Lieferant: Whitehouse Scientific
WHITPS225EA 695 EUR
WHITPS225 WHITPS237 WHITPS202 WHITPS180 WHITPS181 WHITPS205 WHITPS227 WHITPS316 WHITPS194
Sieve standards, polydisperse particle standard
Standards Partikelgrößenanalyse-Standards
The Polydisperse particle standards formally known as the BCR 'Mirror' standards were made both to assess the performance of the analytical laboratories and to augment a limited supply of the official standards. Although sub-divided, the BCR samples were never certified, which leaves these new polydisperse particle standards as the most comprehensively analysed set of spherical particle size reference standards available.
  • Calibration for any method of particle size analysis
  • Unambiguous primary methods used - microscopy, sieving, sedimentation and the Coulter method
Traceability through NIST, BCR and NPL standards.