Dropping bottles, LDPE, Wheaton®
Lieferant: DWK Life Sciences
215-4142EA 209 EUR
215-4142 215-4137 WHEAW242824-A 215-4143 WHEAW242831-A WHEAW242823-A 215-4141 215-4140 215-4136 215-4144 WHEAW242833-A 215-4147 WHEAW242834-A 215-4139 215-4149 WHEAW242835-A WHEAW242836-A 215-4138 215-4145 WHEAW242832-A WHEAW242825-A WHEAW242822-A WHEAW242821-A WHEAW242826-A 215-4148 215-4146
Dropping bottles, LDPE, Wheaton®
Flaschen Tropfflaschen
Dropping bottles made of natural or white LDPE. The white LDPE bottles tends to protect the sensitive samples from UV light.
  • Good chemical resistance
  • Good temperature resistance