Screw caps for high recovery vials, E-Z Ex-Traction™, WHEATON®
Lieferant: DWK Life Sciences
E-Z Ex-Traction™
WHEAW240831EA 1740 EUR
WHEAW240831 WHEAW240842 WHEAW240832 WHEAW240840 WHEAW242710 WHEAW240830 WHEAW240841 WHEAW240846 WHEAW240836 WHEAW240844
Screw caps for high recovery vials, E-Z Ex-Traction™, WHEATON®
Verschlüsse Verschlüsse für Reagenz- und Probenröhrchen
Choice of black phenolic, white polypropylene or urea screw caps
  • PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product
  • Bonded PTFE faced silicone liner ideal for storage of volatile solvents, sensitive compounds and corrosive chemicals