Dropper tips, WHEATON®
Lieferant: DWK Life Sciences
WHEAW242406-AEA 412 EUR
WHEAW242406-A WHEAW242431-01-A WHEAW242408-A WHEAW242402-A WHEAW242404-A WHEA242420 WHEAW242401-01-A WHEA242422 WHEA242421 WHEA242424 WHEA242423 WHEA242426 WHEA242425 WHEA242428 WHEA242427 WHEAW242438-A WHEAW242415-A WHEAW242413-A WHEAW242436-A WHEAW242411-A WHEAW242434-A WHEAW242432-A WHEA242411 WHEA242410 WHEA242413 WHEA242412 WHEA242415 WHEA242414 WHEA242417 WHEA242416 WHEA242418 WHEAW242407-A WHEAW242403-A WHEAW242405-A WHEA242440 WHEAW242420-A WHEAW242401-A WHEA242402 WHEA242401 WHEA242404 WHEA242403 WHEA242406 WHEA242405 WHEAW242411-01-A WHEA242408 WHEA242407 215-4150 WHEAW242418-A WHEAW242416-A WHEAW242437-A WHEAW242414-A WHEAW242433-A WHEAW242412-A WHEA242431 WHEAW242431-A WHEA242430 WHEAW242410-A WHEA242433 WHEAW242440-A WHEA242432 WHEA242434 WHEA242437 WHEA242436 WHEA242438 215-4153 215-4152 215-4151
Dropper tips, WHEATON®
Verschlüsse Tropfverschlüsse
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
Bestellinformation: Tips available in different colours and packaged in case of 100 or 1000.