Screw caps for glass bottles, WHEATON®
Lieferant: DWK Life Sciences
WHEA239209EA 61.9 EUR
WHEA239209 WHEA239290 WHEA239291 WHEA239243 WHEA239287 WHEA239288 WHEA239244 WHEA239241 WHEA239285 WHEA239286 WHEA239242 WHEA239283 WHEA239240 WHEA239284 WHEA239281 WHEA239207 WHEA239401 WHEA239203 WHEA239446 WHEA239289 WHEA239201 WHEA239443 WHEA239246 WHEA239211 WHEA239494 WHEA239292 WHEA239218 WHEA239216 WHEA239217 WHEA239214 WHEA239213 WHEA239222 WHEA239220 WHEA239229 WHEA239425 WHEA239227 WHEA239225 WHEA239226 WHEA239233 WHEA239274 WHEA239275 WHEA239231 WHEA239273 WHEA239238 WHEA239239 WHEA239236 WHEA239237 WHEA239235 WHEA239433
Screw caps for glass bottles, WHEATON®
Verschlüsse Flaschenverschlüsse
Solid top screw caps made of PP with liners. White polypropylene screw cap provides clear surface for writing on cap.
  • Poly-vinyl liner: Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Use these caps with WHEATON or other manufacturers# glass bottles or vials