Sheets for ÄKTA Systems, Benchkote®, Whatman™
Lieferant: Whatman products (Cytiva)
Whatman™ Whatman™ Benchkote®
WHAT2300-10065EA 27 EUR
WHAT2300-10065 WHAT2300-10064 WHAT2300-10074 WHAT2300-10063 WHAT2300-10062 WHAT2300-10073 WHAT2300-10094 WHAT2300-10072 WHAT2300-10061 WHAT2300-10060 WHAT2300-10093 WHAT2300-10092
Sheets for ÄKTA Systems, Benchkote®, Whatman™
Oberflächenschutz Oberflächenschutz
These Benchkote® sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems
Benchkote® material has a high quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.