Gas safety adapter C 206, 1,5 bar for Flame 110
Lieferant: WLD-Tec
WART2.110.800EA 139.5 EUR
WART2.110.800
Gas safety adapter C 206, 1,5 bar for Flame 110
Heizgeräte Gasbrenner
Gas-Safety Adapter C 206, 1,5 bar for Flame 110.
  • For cartridges C 206,190 g from Camping gas
  • Continuous cartridge operation 4,5 h
With integrated pressure regulator 1,5 bar, shut-off valve and DVGW tubing (0,5 m).