Sicherheitsschuhe, zum Schnüren,, Office
Lieferant: Uvex
uvex office
UVEX9542.2/46EA 98.3 EUR
UVEX9542.2/46 UVEX9542.2/45 UVEX9542.2/42 UVEX9542.2/43 UVEX9542.2/47 UVEX9542.2/44 UVEX9542.2/40 UVEX9542.2/39 UVEX9542.2/41 UVEX9542.2/38
Sicherheitsschuhe, zum Schnüren,, Office
Schuhe und Stiefel
Elegant office shoe in brogue style and wing tip made from supple, full-grain leather upper and a Polyurethane sole (cut and abrasion resistant, heat-resistant for brief exposure up to +120 °C; oil and petrol-resistant).
  • Breathable lining
  • Ultra comfortable insole
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Standards: EN ISO 20345: 2011; S1P, SRA