ESD safety ankle boots, lace-up, uvex 1, 8532
Lieferant: Uvex
UVEX8532.9/43EA 97.3 EUR
UVEX8532.9/43 UVEX8532.9/42 UVEX8532.9/45 UVEX8532.9/44
ESD safety ankle boots, lace-up, uvex 1, 8532
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PU sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/blue
Width: 12
Standards: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC