ESD safety shoes, lace-up, uvex 2 trend, 6949
Lieferant: Uvex
uvex 2 trend
UVEX6949.8/49EA 73.1 EUR
UVEX6949.8/49 UVEX6949.7/38 UVEX6949.9/39 UVEX6949.9/38 UVEX6949.8/47 UVEX6949.8/48 UVEX6949.8/45 UVEX6949.8/46 UVEX6949.8/43 UVEX6949.8/44 UVEX6949.8/41 UVEX6949.7/51 UVEX6949.8/42 UVEX6949.7/52 UVEX6949.9/52 UVEX6949.8/40 UVEX6949.9/51 UVEX6949.7/50 UVEX6949.9/50 UVEX6949.7/39 UVEX6949.7/48 UVEX6949.8/38 UVEX6949.8/39 UVEX6949.7/49 UVEX6949.9/49 UVEX6949.7/46 UVEX6949.7/47 UVEX6949.9/48 UVEX6949.9/47 UVEX6949.7/44 UVEX6949.7/45 UVEX6949.9/46 UVEX6949.9/45 UVEX6949.7/42 UVEX6949.9/44 UVEX6949.7/43 UVEX6949.8/52 UVEX6949.9/43 UVEX6949.7/40 UVEX6949.7/41 UVEX6949.9/42 UVEX6949.9/41 UVEX6949.8/50 UVEX6949.8/51 UVEX6949.9/40
ESD safety shoes, lace-up, uvex 2 trend, 6949
Schuhe und Stiefel
Sporty S2 safety shoe with wide fit.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Reflective details for enhanced visibility
  • Breathable, water repellent microvelour
  • Steel toe cap
  • Shock absorbing heel and forefoot
  • Climazone technology helps maintain foot temperature for optimum wearer comfort
Safety Shoes feature an ergonomically designed outsole that offers excellent slip resistance, as well as a stabilising uvex anti-twist heel cap designed to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances. Soft wearer feeling provides comfortable padding on collar and dust tongue. Removable anti-static comfortable insole, with a moisture transport system.
Colour: Black/grey
Standards: EN 61340 (ESD); EN ISO 20345:2011 S2 SRC; certified with uvex medicare.