ESD safety ankle boots, lace-up, uvex 2 trend, S2, 6935
Lieferant: Uvex
UVEX6935.9/48EA 74.8 EUR
UVEX6935.9/48 UVEX6935.7/43 UVEX6935.8/47 UVEX6935.8/45 UVEX6935.8/43 UVEX6935.9/39 UVEX6935.7/40 UVEX6935.8/44 UVEX6935.9/46 UVEX6935.7/45 UVEX6935.7/49 UVEX6935.8/46 UVEX6935.9/43 UVEX6935.9/40 UVEX6935.9/38 UVEX6935.7/42 UVEX6935.9/50 UVEX6935.8/39 UVEX6935.7/50 UVEX6935.9/44 UVEX6935.7/51 UVEX6935.9/52 UVEX6935.9/51 UVEX6935.7/38 UVEX6935.8/41 UVEX6935.9/45 UVEX6935.8/51 UVEX6935.9/47 UVEX6935.8/50 UVEX6935.7/48 UVEX6935.7/52 UVEX6935.8/42 UVEX6935.8/40 UVEX6935.8/49 UVEX6935.7/41 UVEX6935.9/49 UVEX6935.8/48 UVEX6935.9/42 UVEX6935.7/39 UVEX6935.9/41 UVEX6935.7/46 UVEX6935.8/52 UVEX6935.7/47 UVEX6935.8/38 UVEX6935.7/44
ESD safety ankle boots, lace-up, uvex 2 trend, S2, 6935
Schuhe und Stiefel
Sporty S2 safety boot with wide fit.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Water repellent leather
  • Steel toecap
  • Smooth-running, dirt and heat-resistant lacing system
  • Reflective details for enhanced visibility
Safety boots feature an ergonomically designed outsole with excellent slip resistance. A stabilising uvex anti-twist heel cap is designed to prevent ankle twisting. All sole materials are free of silicones, plasticisers and other paint wetting impairment substances.
For medium-duty applications.
Colour: Blue/black
Standards: EN 61340 (ESD); EN ISO 20345:2011; S2, SRC