ESD safety shoes, lace-up, uvex 2 trend, 6934, S2
Lieferant: Uvex
UVEX6934.9/44EA 73 EUR
UVEX6934.9/44 UVEX6934.8/51 UVEX6934.7/41 UVEX6934.7/42 UVEX6934.7/47 UVEX6934.8/52 UVEX6934.9/52 UVEX6934.9/42 UVEX6934.8/49 UVEX6934.7/44 UVEX6934.8/47 UVEX6934.8/48 UVEX6934.8/44 UVEX6934.7/43 UVEX6934.9/47 UVEX6934.7/38 UVEX6934.9/38 UVEX6934.7/48 UVEX6934.7/49 UVEX6934.8/45 UVEX6934.9/45 UVEX6934.8/42 UVEX6934.9/39 UVEX6934.9/46 UVEX6934.9/41 UVEX6934.9/40 UVEX6934.7/45 UVEX6934.8/46 UVEX6934.8/39 UVEX6934.7/40 UVEX6934.8/38 UVEX6934.8/41 UVEX6934.9/49 UVEX6934.9/50 UVEX6934.7/46 UVEX6934.9/48 UVEX6934.8/40 UVEX6934.7/39 UVEX6934.8/50 UVEX6934.7/52 UVEX6934.9/51 UVEX6934.7/51 UVEX6934.9/43 UVEX6934.8/43 UVEX6934.7/50
ESD safety shoes, lace-up, uvex 2 trend, 6934, S2
Schuhe und Stiefel
Sporty S2 safety shoe with wide fit.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Ergonomically designed outsole offers excellent slip resistance
  • Water repellent leather
  • Smooth-running, dirt- and heat-resistant lacing system
  • Reflective details for enhanced visibility
Safety Shoes feature a penetration resistant steel midsole (S3 models) and steel toe cap. Stabilising uvex anti-twist heel cap is designed to prevent ankle twisting. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/blue
Standards: EN 61340 (ESD); EN ISO 20345:2011, S2, SRC