ESD safety shoes, lace-up, uvex 2 trend, 6907
Lieferant: Uvex
uvex 2
UVEX6907.9/39EA 67.8 EUR
UVEX6907.9/39 UVEX6907.8/49 UVEX6907.8/45 UVEX6907.7/39 UVEX6907.8/46 UVEX6907.7/38 UVEX6907.8/47 UVEX6907.8/48 UVEX6907.9/38 UVEX6907.8/41 UVEX6907.7/50 UVEX6907.8/42 UVEX6907.8/43 UVEX6907.8/44 UVEX6907.9/50 UVEX6907.7/52 UVEX6907.9/51 UVEX6907.7/51 UVEX6907.8/40 UVEX6907.9/52 UVEX6907.7/47 UVEX6907.8/38 UVEX6907.8/39 UVEX6907.7/46 UVEX6907.7/45 UVEX6907.7/44 UVEX6907.9/46 UVEX6907.9/47 UVEX6907.7/49 UVEX6907.9/48 UVEX6907.9/49 UVEX6907.7/48 UVEX6907.8/52 UVEX6907.9/42 UVEX6907.9/43 UVEX6907.9/44 UVEX6907.9/45 UVEX6907.7/43 UVEX6907.7/42 UVEX6907.9/40 UVEX6907.8/50 UVEX6907.7/41 UVEX6907.7/40 UVEX6907.9/41 UVEX6907.8/51
ESD safety shoes, lace-up, uvex 2 trend, 6907
Schuhe und Stiefel
ESD sporty S1 safety shoe for men with wide fit and slip resistance PU sole.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable, perforated microvelours
  • Steel toe cap
  • Reflective details for enhanced visibility
  • Excellent shock absorption
  • Uvex climazone foot climate management, uvex 1 climate comfort insole, uvex multi-width system helps maintain foot temperature for optimum wearer comfort
Safety Shoes feature an ergonomically designed outsole with excellent slip resistance, as well as a stabilising uvex anti-twist heel cap designed to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
The high-quality outsole materials, sports influenced design and bold colours with maximum comfort and reliable safety. Medium-duty applications. Lining material: distance mesh.
Colour: Black/grey/orange
Standards: EN 61340 (ESD); EN ISO 20345:2011, S1, SRC; certified with uvex medicare