ESD safety shoes, lace-up, uvex 1 sport, 6593
Lieferant: Uvex
uvex 1 sport
UVEX6593.7/35EA 98.4 EUR
UVEX6593.7/35 UVEX6593.8/48 UVEX6593.8/49 UVEX6593.8/44 UVEX6593.7/39 UVEX6593.8/45 UVEX6593.7/38 UVEX6593.8/46 UVEX6593.7/37 UVEX6593.8/47 UVEX6593.7/36 UVEX6593.8/40 UVEX6593.8/41 UVEX6593.8/42 UVEX6593.8/43 UVEX6593.7/52 UVEX6593.7/51 UVEX6593.7/50 UVEX6593.7/46 UVEX6593.8/37 UVEX6593.7/45 UVEX6593.8/38 UVEX6593.8/39 UVEX6593.7/44 UVEX6593.7/43 UVEX6593.7/49 UVEX6593.7/48 UVEX6593.8/35 UVEX6593.7/47 UVEX6593.8/36 UVEX6593.8/51 UVEX6593.8/52 UVEX6593.7/42 UVEX6593.7/41 UVEX6593.7/40 UVEX6593.8/50
ESD safety shoes, lace-up, uvex 1 sport, 6593
Schuhe und Stiefel
The uvex 1 sport is both youthful and stylish combined with first-class wearer comfort, thanks to the lightweight design, climate-optimised high-tech materials, and ergonomically designed outsole.
  • Work shoes meet the ESD specifications with leakage resistance under 35 megaohms
  • Breathable textile upper
  • No toe cap
This modern, ultra-lightweight and flexible work shoe is made from synthetic materials, so it is suitable for people allergic to chrome. Sole materials are free from silicones, plasticisers, and other substances that interfere with wetting agents. The ergonomically designed outsole is made from dual-density polyurethane that offers excellent slip resistance.
Colour: Black
Standards: Protection class O1 FO SRC in accordance with EN ISO 20347:2012; EN 61340 (ESD).