ESD safety shoes, lace-up, 6556, uvex 1
Lieferant: Uvex
uvex 1
UVEX6556.9/46EA 93.8 EUR
UVEX6556.9/46 UVEX6556.9/47 UVEX6556.9/44 UVEX6556.8/35 UVEX6556.9/45 UVEX6556.8/37 UVEX6556.8/36 UVEX6556.9/48 UVEX6556.8/39 UVEX6556.9/49 UVEX6556.8/38 UVEX6556.9/42 UVEX6556.9/43 UVEX6556.9/40 UVEX6556.9/41 UVEX6556.8/44 UVEX6556.8/43 UVEX6556.8/46 UVEX6556.8/45 UVEX6556.8/48 UVEX6556.7/36 UVEX6556.8/47 UVEX6556.7/35 UVEX6556.7/38 UVEX6556.8/49 UVEX6556.7/37 UVEX6556.7/39 UVEX6556.9/50 UVEX6556.8/40 UVEX6556.8/42 UVEX6556.9/51 UVEX6556.9/52 UVEX6556.8/41 UVEX6556.7/43 UVEX6556.7/42 UVEX6556.7/45 UVEX6556.7/44 UVEX6556.7/47 UVEX6556.7/46 UVEX6556.7/49 UVEX6556.7/48 UVEX6556.8/51 UVEX6556.8/50 UVEX6556.7/41 UVEX6556.8/52 UVEX6556.7/40 UVEX6556.9/35 UVEX6556.9/36 UVEX6556.9/39 UVEX6556.9/37 UVEX6556.9/38 UVEX6556.7/50 UVEX6556.7/52 UVEX6556.7/51
ESD safety shoes, lace-up, 6556, uvex 1
Schuhe und Stiefel
Sporty, multifunctional safety shoe available in multiple widths.
  • Breathable, perforated, high-tech microvelour upper material
  • Elastic laces for quick, individual adjustment
  • Virtually seamless design to eliminate pressure points
  • Free of lacquer wetting inhibitors such as silicones and phthalates
  • Ideal shape around the joints for optimal torsion and good flexibility
  • Joint insert for climbing ladders securely
  • Pivot point eases turning when standing
These especially lightweight and flexible safety shoes feature a multi-layer-shock absorption system using dual density polyurethane for walking comfort. Additional features include self-cleaning tread optimised for slip resistance, uvex Climazone technology for optimum wearer comfort, and distance mesh lining for optimum breathability. The latest generation of 100% metal-free uvex xenova® toe cap offers compact design, anatomical shape, good side stability, no thermal conductivity, and anti-magnetic properties.
Suitable for people allergic to chrome, as they are made from synthetic materials.
Fulfils ESD requirements: resistance to ground inferior to 35 megaohms.
Colour: Black/silver
Standards: EN 61340 (ESD); EN ISO 20345:2011 S1 SRC