Chemical resistant gloves, uvex profabutyl B-05R
Lieferant: Uvex
uvex profabutyl
UVEX6094909EA 64.8 EUR
UVEX6094909 UVEX6094908 UVEX6094910 UVEX6094911 UVEX6094907
Chemical resistant gloves, uvex profabutyl B-05R
Handschuhe
These unsupported gloves made from 100% butyl rubber provide a good resistance to polar compounds such as esters, ketones, aldehydes, amines and saturated saline solutions, plus acids and alkalis.
  • Seamlessly coated with bromobutyl (approx. 0,50 mm)
  • Good grip in damp and wet areas
  • Highly flexible, even at low temperatures
  • Impermeable to water vapour, gases and toxic substances
Please note: Butyl is not resistant to oil, grease, aliphatic and aromatic hydrocarbons or chlorinated hydrocarbons.
Standards: EN 388:2016 (2010X), EN ISO 374-1:2016 Type A (ABIKLP)
Zubehörinformationen: EN 388:2016 (2010X), EN ISO 374-1:2016 Type A (ABIKLP)