Anti-Transglutaminase Rabbit Polyclonal Antibody
Lieferant: United States Biological
USBIT3627-01FEA 535 EUR
USBIT3627-01F
Anti-Transglutaminase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Transglutaminase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human