Anti-SH2 domain protein 2A Sheep Polyclonal Antibody
Lieferant: United States Biological
USBIS1010-90KEA 796 EUR
USBIS1010-90K
Anti-SH2 domain protein 2A Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: SH2 domain protein 2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human