Anti-Scavenger Receptor A Type I/II Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A458]

Lieferant: United States Biological
USBIS0099-06EA 405 EUR
USBIS0099-06
Anti-Scavenger Receptor A Type I/II Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A458]
Antikörper
Recommended dilution: Flow Cytometry:

Type: Primary
Antigen: Scavenger Receptor A Type I/II
Clonality: Monoclonal
Clone: 6A458
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR