Anti-Phosphatase, Alkaline, Human, Bone Sheep Polyclonal Antibody

Lieferant: United States Biological
USBIP4071-11BEA 570 EUR
USBIP4071-11B
Anti-Phosphatase, Alkaline, Human, Bone Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1500-1:15000

Type: Primary
Antigen: Phosphatase, Alkaline, Human, Bone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR