Anti-PMAIP1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIN5376-80EEA 574 EUR
USBIN5376-80E
Anti-PMAIP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PMAIP1 (phorbol-12-myristate-13-acetate-induced protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR