Anti-MCAF Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIM2690-10BEA 450 EUR
USBIM2690-10B
Anti-MCAF Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:2000

Type: Primary
Antigen: MCAF
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR