Anti-LTF Goat Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIL1021-82BEA 580 EUR
USBIL1021-82B
Anti-LTF Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:2000-1:10000

Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR