Anti-IgY heavy chain Mouse Monoclonal Antibody [clone: 7E138]
Lieferant: United States Biological
USBII1906-04EEA 608 EUR
USBII1906-04E
Anti-IgY heavy chain Mouse Monoclonal Antibody [clone: 7E138]
Antikörper
Type: Secondary
Antigen: IgY, Heavy Chain
Clonality: monoclonal
Clone: 7E138
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen