Anti-IgG Donkey Polyclonal Antibody (Biotin)
Lieferant: United States Biological
USBII1903-89F2EA 427 EUR
USBII1903-89F2
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse