Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 10B936/3H814]
Lieferant: United States Biological
USBIC2269-94AEA 561 EUR
USBIC2269-94A
Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 10B936/3H814]
Antikörper
Do not freeze!
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: Monoclonal
Clone: 10B936/3H814
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mixed
Reactivity:
Bestellen