Anti-IGFBP1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI143592-50EA 425 EUR
USBI143592-50 USBI143592
Anti-IGFBP1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.25-1 ?g/ml

Type: Primary
Antigen: IGFBP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR