Anti-PLP1 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI131489EA 565 EUR
USBI131489
Anti-PLP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLP1 (proteolipid protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR