Anti-IDH2 Mouse Monoclonal Antibody [clone: 5F11]
Lieferant: US Biological
USBI128208EA 522 EUR
USBI128208
Anti-IDH2 Mouse Monoclonal Antibody [clone: 5F11]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: IDH2 (isocitrate dehydrogenase 2 (NADP+), mitochondrial)
Clonality: monoclonal
Clone: 5F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human