Anti-FCER2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI124582EA 565 EUR
USBI124582
Anti-FCER2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FCER2 (Fc fragment of IgE, low affinity II, receptor for (CD23))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR