Anti-ACSS1 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI122909EA 522 EUR
USBI122909
Anti-ACSS1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACSS1 (acyl-CoA synthetase short-chain family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR