Anti-ABI2 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI122825EA 522 EUR
USBI122825
Anti-ABI2 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ABI2 (abl-interactor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR