Anti-SLC25A37 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: United States Biological
USBI041869-APEA 852 EUR
USBI041869-AP
Anti-SLC25A37 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: SLC25A37 (solute carrier family 25 (mitochondrial iron transporter), member 37)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR