Anti-RBMS2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI040904EA 587 EUR
USBI040904
Anti-RBMS2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: RBMS2 (RNA binding motif, single stranded interacting protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR