Anti-MYO19 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
USBI038743-BIOTINEA 852 EUR
USBI038743-BIOTIN
Anti-MYO19 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Type: Primary
Antigen: MYO19 (myosin XIX)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human