Anti-CACNA2D2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
USBI033127-PEEA 852 EUR
USBI033127-PE
Anti-CACNA2D2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: CACNA2D2 (calcium channel, voltage-dependent, alpha 2/delta subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse