Size exclusion chromatography sorbents, Toyopearl®
Lieferant: Tosoh Bioscience
TOYO0014691EA 1410 EUR
TOYO0014691 TOYO0019820 TOYO0019821 TOYO0014690 TOYO0007459 TOYO0007467 TOYO0007466 TOYO0007469 TOYO0007465 TOYO0014688 TOYO0019816 TOYO0014689 TOYO0014686 TOYO0014687 TOYO0019819 TOYO0019812 TOYO0019813 TOYO0019814 TOYO0019815 TOYO0014681 TOYO0019810 TOYO0019811 TOYO0014684 TOYO0014685 TOYO0014682 TOYO0014683 TOYO0007449 TOYO0007448 TOYO0007455 TOYO0007457 TOYO0007451 TOYO0007453 TOYO0021481 TOYO0019807 TOYO0019808
Size exclusion chromatography sorbents, Toyopearl®
Chromatographie-Sorbentien HPLC-Sorbentien
Toyopearl resins can be used in organic solvents or mixtures of organic solvents and water. The compatibility of DMF with Toyopearl also permits the separation of hydrophobic substances such as polystyrenes.
  • Removal of surfactants such as Triton® X-100 from biological solutions by an adsorption mechanism
  • Use in hydrophobic interaction chromatography (HIC) for the separation of very hydrophobic molecules
  • Suitable for HIC separations as a guard column for hydrophobic impurities
  • Used as a stationary phase for either normal or reversed phase separations depending on solvent system selected
The Toyopearl HW-type resins are commonly used in size exclusion chromatography and desalting applications.