Multi-position magnetic stirrers, MIXdrive MTP series
Lieferant: 2mag
MIX
TMAG90250EA 720 EUR
TMAG90250 TMAG43012 TMAG43024 TMAG43006 TMAG90252 TMAG43096
Multi-position magnetic stirrers, MIXdrive MTP series
Rührer Mehrstellen-Magnetrührer
Multi-position magnetic stirrers with hermetically sealed stainless steel housing which is water-, germ- and dust-proof. Suitable for shaking micotitre plates in CO2 incubators and ovens.
  • Maintenance- and wear-free inductive drive
  • 100% synchronised jerk-free mixing
  • Submersible in water and oil baths
MIXcontrol MTP is an external control unit made from easy to clean, resistant stainless steel housing. It includes SoftStart for reliable and safe acceleration of the stirring bar and QuickSet for setting of start and maximum speed. It has automatic memory function to save last setting and clear digital display for speed and power setting. It provide jerk-free stirring at low speed and power setting at four steps.
Standards: IP protection class: IP 68
Bestellinformation: External control unit (MIXcontrol MTP) must be ordered separately.