Probes for testo 440
Lieferant: Testo
TEST0615.5505EA 45 EUR
TEST0615.5505 TEST0635.9370 620-2660 620-2662 TEST0635.9430 TEST0636.9732 TEST0635.9431 620-2661 TEST0635.9432 TEST0635.9532 TEST0635.9570 TEST0635.9372 TEST0635.9571 TEST0635.9572 TEST0635.1570 TEST0632.1271 620-2659 TEST0632.1272 620-2658 TEST0635.1032 TEST0635.1572 TEST0632.1270 TEST0636.9770 TEST0632.1550 TEST0636.9772 TEST0636.9775 TEST0636.9731 TEST0636.9730 TEST0632.1552 TEST0615.1212 TEST0615.1712 TEST0635.0551 TEST0628.0152
Probes for testo 440
Temperaturfühler
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.