Gas leak detector
Lieferant: Testo
TEST0632.3162EA 321 EUR
TEST0632.3162
Gas leak detector
Lecksuchsysteme Gas-Lecksuchsysteme
This gas leak detector is designed for fast checking measurements.
  • Trend display shows maximum leakage
  • Optical presentation of the gas concentrations
  • Flexible measurement probe for inaccessible places
  • Earphone connection for secure leakage localisation in loud surroundings
Lieferumfang: Supplied with measurement probe, rechargeable battery and earphones.