Gas leak detector
Lieferant: Testo
TEST0632.0316EA 215 EUR
TEST0632.0316
Gas leak detector
Lecksuchsysteme Gas-Lecksuchsysteme
This gas leak monitoring instrument is specially suitable for use on methane.
  • Flexible measurement probe
  • For hard to reach locations
  • Optical and audible alarms when limits are exceeded
Lieferumfang: Supplied with instruction manual and battery.