Brushes for tubes and barrels
Lieferant: Socorex
SOCO1.170.305EA 33.4 EUR
SOCO1.170.305 SOCO1.170.300 SOCO1.170.320
Brushes for tubes and barrels
Bürsten
Cylinder and tube brushes with tufted end and plastic handle for glass barrel and tube cleaning.
Lieferumfang: The set includes: Two brushes for 0,3/0,5/1 ml barrels, two brushes fpr 1/2/5 ml barrels and two brushes for 5/10/20 ml barrels.