Propyltriethoxysilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS14100.100EA 115 EUR
SILNSILS14100.100 SILNSILS14100.25
Propyltriethoxysilan
Propyltriethoxysilan
Formel: C₉H₂₂O₃Si
Molecular Weight: 206,36 g/mol
MDL: MFCD00026756
CAS-Nummer: 2550-02-9
EINECS: 219-842-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR