Inosin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
SIALI4125-10GEA
107
EUR
SIALI4125-10G
SIALI4125-25G
SIALI4125-100G
SIALI4125-1G
PFIZI-4125
Inosin, Sigma-Aldrich®
Inosin
Formel:
C₁₀H₁₂N₄O₅ Molecular Weight: 268,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066770 CAS-Nummer: 58-63-9 EINECS: 200-390-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...